KEY001124629 Key Organics / BIONET

3-amino-4-[(4-methylphenyl)sulfanyl]benzonitrile

  • MDL number MFCD00793310
  • Cas Number : 337924-03-5
  • Molecular Weight : 240.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 101
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1608

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10K-614S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart