KEY001124656 Key Organics / BIONET

4-chloro-2-(trifluoromethyl)quinazoline

  • MDL number MFCD00793683
  • Cas Number : 52353-35-2
  • Molecular Weight : 232.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65 - 68
Purity 95
Storage temp. Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1635

Safety Information

signalword Warning
pcodes P232,P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10L-019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart