KEY001124673 Key Organics / BIONET

methyl 2-(4-chlorophenyl)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD00792936
  • Cas Number : 337924-65-9
  • Molecular Weight : 267.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138 - 140
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1652

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10L-037_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart