KEY001124790 Key Organics / BIONET

5-[(4-methoxyphenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00975682
  • Cas Number : 321998-19-0
  • Molecular Weight : 324.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94 - 98
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1791

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10L-555S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart