KEY001124941 Key Organics / BIONET

2,4-dioxo-1-[(pyridin-4-yl)methyl]-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD00974184
  • Cas Number : 341966-23-2
  • Molecular Weight : 228.211

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 313 - 315
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1952

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10L-933_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart