KEY001125145 Key Organics / BIONET

(Z)-4-[(2-chloro-1,3-thiazol-5-yl)methoxy]-N'-hydroxybenzene-1-carboximidamide

  • MDL number MFCD01568715
  • Cas Number : 341967-76-8
  • Molecular Weight : 283.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179 - 181
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2207

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10N-071_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart