KEY001125281 Key Organics / BIONET

3,3'-dichloro-5,5'-bis(trifluoromethyl)-2H-[1,2'-bipyridine]-2-one

  • MDL number MFCD01568202
  • Cas Number : 96741-18-3
  • Molecular Weight : 377.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107 - 109
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2358

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10N-740_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart