KEY001125455 Key Organics / BIONET

1-phenyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD01749393
  • Cas Number : 116834-08-3
  • Molecular Weight : 253.261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185 - 189
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2548

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10P-383S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart