KEY001125610 Key Organics / BIONET

2-(1H-imidazol-1-yl)benzamide

  • MDL number MFCD00127076
  • Cas Number : 25373-52-8
  • Molecular Weight : 187.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176 - 178
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3727

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11A-032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart