KEY001125646 Key Organics / BIONET

methyl 2-(4-chlorophenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylate

  • MDL number MFCD00140323
  • Cas Number : 446276-02-4
  • Molecular Weight : 313.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152 - 154
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3776

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11B-030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart