KEY001125686 Key Organics / BIONET

methyl 1-oxo-2-[(pyridin-3-yl)methyl]-1,2-dihydroisoquinoline-4-carboxylate

  • MDL number MFCD00140334
  • Cas Number : 446276-03-5
  • Molecular Weight : 294.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164 - 166
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3822

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11B-128_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart