KEY001125924 Key Organics / BIONET

3-methyl-1-phenyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD00140640
  • Cas Number : 321385-93-7
  • Molecular Weight : 317.315

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104 - 105
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4084

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11E-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart