KEY001125981 Key Organics / BIONET

ethyl 2-cyano-2-(1,3-dioxolan-2-ylidene)acetate

  • MDL number MFCD00172357
  • Cas Number : 121020-70-0
  • Molecular Weight : 183.163

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 140
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4176

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11E-918_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart