KEY001125982 Key Organics / BIONET

2-amino-4-(2-hydroxyethoxy)-6-oxo-1,6-dihydropyrimidine-5-carbonitrile

  • MDL number MFCD00172358
  • Cas Number : 126865-37-0
  • Molecular Weight : 196.166

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 278 - 280
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4177

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11E-920_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart