KEY001126018 Key Organics / BIONET

methyl 3-(5-cyano-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)thiophene-2-carboxylate

  • MDL number MFCD00172924
  • Cas Number : 446276-11-5
  • Molecular Weight : 277.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260 - 262
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4215

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11E-961_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart