KEY001126019 Key Organics / BIONET

ethyl 5-amino-3-(2-hydroxyethoxy)-1,2-oxazole-4-carboxylate

  • MDL number MFCD00172362
  • Cas Number : 446276-12-6
  • Molecular Weight : 216.193

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128 - 130
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4216

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11E-962_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart