KEY001126053 Key Organics / BIONET

2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoic acid

  • MDL number MFCD00202110
  • Cas Number : 303148-48-3
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 147 - 150
Iupac Name 2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoic acid
Inchi Code 1S/C6H9N3O2/c1-6(2,5(10)11)9-4-7-3-8-9/h3-4H,1-2H3,(H,10,11)
Inchi Key XCYWDFDMXREMCT-UHFFFAOYSA-N

Description

Melting Point 147 - 150
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4257

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11F-063_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart