KEY001126135 Key Organics / BIONET

5-(4-chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00215390
  • Cas Number : 109925-13-5
  • Molecular Weight : 250.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74 - 75
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4360

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11F-431S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart