KEY001126158 Key Organics / BIONET

2-(prop-2-en-1-yloxy)naphthalene-1-carbaldehyde

  • MDL number MFCD00231645
  • Cas Number : 19530-43-9
  • Molecular Weight : 212.248

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81 - 83
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4385

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11F-907_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart