KEY001126165 Key Organics / BIONET

2-(2-phenyl-1,3-thiazol-4-yl)benzoic acid

  • MDL number MFCD00141945
  • Cas Number : 60510-56-7
  • Molecular Weight : 281.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162 - 164
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4392

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11F-915_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart