KEY001126198 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-N-(2-hydroxyethyl)piperidine-4-carboxamide

  • MDL number MFCD00172258
  • Cas Number : 303150-23-4
  • Molecular Weight : 351.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174 - 176
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4426

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11F-955_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart