KEY001126200 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-N-(2-hydroxyethyl)-N-methylpiperidine-4-carboxamide

  • MDL number MFCD00172259
  • Cas Number : 303150-26-7
  • Molecular Weight : 365.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113 - 115
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4427

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11F-957_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart