KEY001126236 Key Organics / BIONET

3-(prop-2-yn-1-ylsulfanyl)-1H-1,2,4-triazol-5-amine

  • MDL number MFCD00141528
  • Cas Number : 53918-44-8
  • Molecular Weight : 154.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121 - 123
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4463

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11G-051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart