KEY001126261 Key Organics / BIONET

3-[(carbamoylmethyl)sulfanyl]thiophene-2-carboxylic acid

  • MDL number MFCD00663911
  • Cas Number : 303150-84-7
  • Molecular Weight : 217.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231 - 235
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4497

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11G-342S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart