KEY001126377 Key Organics / BIONET

4-hydroxy-6-(4-methylphenyl)-2-phenylpyrimidine-5-carbonitrile

  • MDL number MFCD00794098
  • Cas Number : 477853-90-0
  • Molecular Weight : 287.322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 350 - 353
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4661

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11H-032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart