KEY001126383 Key Organics / BIONET

4-(4-fluorophenyl)-6-hydroxy-2-phenylpyrimidine-5-carbonitrile

  • MDL number MFCD00794103
  • Cas Number : 477853-95-5
  • Molecular Weight : 291.285

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 330
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4667

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11H-038_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart