KEY001126388 Key Organics / BIONET

4-hydroxy-6-(2-methoxyphenyl)-2-phenylpyrimidine-5-carbonitrile

  • MDL number MFCD00794104
  • Cas Number : 477853-96-6
  • Molecular Weight : 303.321

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 277 - 280
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4675

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11H-049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart