KEY001126490 Key Organics / BIONET

(2E)-2-cyano-3-(dimethylamino)but-2-enamide

  • MDL number MFCD00663777
  • Cas Number : 62321-92-0
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 194 - 196
Iupac Name (2E)-2-cyano-3-(dimethylamino)-2-butenamide
Inchi Code 1S/C7H11N3O/c1-5(10(2)3)6(4-8)7(9)11/h1-3H3,(H2,9,11)/b6-5+
Inchi Key CPNKUDCDGKOECR-AATRIKPKSA-N

Description

Melting Point 194 - 196
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/4784

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11H-902_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart