KEY001126946 Key Organics / BIONET

ethyl 2-({[3-(hydroxymethyl)-8-methoxy-3,4-dihydro-2H-1-benzopyran-4-yl](methyl)carbamothioyl}amino)acetate

  • MDL number MFCD00243893
  • Cas Number : 303148-56-3
  • Molecular Weight : 368.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151 - 153
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5320

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11K-925_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart