KEY001127076 Key Organics / BIONET

1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethan-1-one

  • MDL number MFCD00662755
  • Cas Number : 37091-33-1
  • Molecular Weight : 221.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148 - 149
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5472

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11L-568S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart