KEY001127302 Key Organics / BIONET

ethyl 1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD02083398
  • Cas Number : 111493-74-4
  • Molecular Weight : 222.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 60 - 61
Iupac Name ethyl 1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Inchi Code 1S/C8H9F3N2O2/c1-3-15-7(14)5-4-13(2)12-6(5)8(9,10)11/h4H,3H2,1-2H3
Inchi Key ZZEXDJGNURSJOF-UHFFFAOYSA-N

Description

Melting Point 60 - 61
Purity 95%
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5715

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11M-320S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart