KEY001127387 Key Organics / BIONET

4-(4-fluorophenyl)pyrimidine-2-thiol

  • MDL number MFCD01315002
  • Cas Number : 155957-43-0
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5813

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11M-725_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart