KEY001127430 Key Organics / BIONET

2-(4-benzylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD01568829
  • Cas Number : 303987-22-6
  • Molecular Weight : 287.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107 - 109
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5860

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11N-024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart