KEY001127485 Key Organics / BIONET

2-(4-phenylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD01568830
  • Cas Number : 303987-39-5
  • Molecular Weight : 273.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 119
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/5915

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11N-091_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart