KEY001127599 Key Organics / BIONET

methyl 4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate

  • MDL number MFCD01569173
  • Cas Number : 303987-90-8
  • Molecular Weight : 237.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139 - 141
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6044

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11N-524S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart