KEY001127666 Key Organics / BIONET

5-(benzylsulfanyl)-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD01568740
  • Cas Number : 4858-36-0
  • Molecular Weight : 240.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137 - 139
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6113

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11N-703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart