KEY001127696 Key Organics / BIONET

ethyl 2-[(2-chloro-6-fluorophenyl)methyl]-3-oxobutanoate

  • MDL number MFCD00052428
  • Cas Number : 477856-17-0
  • Molecular Weight : 272.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 @ 0.1mBar
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6138

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11N-747_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart