KEY001127753 Key Organics / BIONET

4-[4-(methoxycarbonyl)piperidin-1-yl]-4-oxobutanoic acid

  • MDL number MFCD01568756
  • Cas Number : 303994-57-2
  • Molecular Weight : 243.259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91 - 93
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6192

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11N-923_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart