KEY001128025 Key Organics / BIONET

2-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)-3-methylbutanoic acid

  • MDL number MFCD02081981
  • Cas Number : 477858-29-0
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 148
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6515

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11P-621_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart