KEY001128042 Key Organics / BIONET

4-methyl-2-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)pentanoic acid

  • MDL number MFCD02082683
  • Cas Number : 298700-70-6
  • Molecular Weight : 247.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185 - 188
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/6535

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11P-650_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart