KEY001128136 Key Organics / BIONET

2-amino-4-tert-butyl-1,3-thiazole-5-carbonitrile

  • MDL number MFCD00214700
  • Cas Number : 303994-99-2
  • Molecular Weight : 181.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189 - 191
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7457

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12A-067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart