KEY001128237 Key Organics / BIONET

1-[5-(2,2,2-trichloroacetyl)-1H-pyrrol-3-yl]propan-1-one

  • MDL number MFCD00141837
  • Cas Number : 111468-90-7
  • Molecular Weight : 268.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190 - 192
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7571

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12C-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart