KEY001128247 Key Organics / BIONET

1-phenyl-1H-1,2,4-triazol-3-ol

  • MDL number MFCD00138713
  • Cas Number : 4231-68-9
  • Molecular Weight : 161.164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 277 - 280
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7581

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12C-058_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart