KEY001128252 Key Organics / BIONET

methyl 5-(2-methoxy-2-oxoethyl)-1-(4-methoxyphenyl)-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD00138716
  • Cas Number : 91306-62-6
  • Molecular Weight : 305.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 82
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7586

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12C-073_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart