KEY001128669 Key Organics / BIONET

1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD00173066
  • Cas Number : 103058-60-2
  • Molecular Weight : 237.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 192 - 194
Iupac Name 1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid
Inchi Code 1S/C10H8ClN3O2/c1-6-12-9(10(15)16)13-14(6)8-4-2-7(11)3-5-8/h2-5H,1H3,(H,15,16)
Inchi Key BVBOJSLEUHHRNY-UHFFFAOYSA-N

Description

Melting Point 192 - 194
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8068

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/12G-370S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart