KEY001129155 Key Organics / BIONET

4-chloro-2-(3-chlorophenyl)-5-[(2-hydroxyethyl)amino]-2,3-dihydropyridazin-3-one

  • MDL number MFCD00793818
  • Cas Number : 320421-79-2
  • Molecular Weight : 300.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151 - 153
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8633

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12K-921_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart