KEY001129240 Key Organics / BIONET

1-methyl-3-(trifluoromethyl)-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

  • MDL number MFCD01814674
  • Cas Number : 338982-15-3
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255 - 258
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8725

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12L-521S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart