KEY001129362 Key Organics / BIONET

1-(2-fluorophenyl)-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD01443629
  • Cas Number : 132407-65-9
  • Molecular Weight : 189.189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 125
Purity 97
Storage temp. RT
Physical Form Off-white to Dark Green Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8861

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/12M-041_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart