KEY001129442 Key Organics / BIONET

4-(4-methylphenyl)pyrimidine-2-thiol

  • MDL number MFCD01315005
  • Cas Number : 477859-71-5
  • Molecular Weight : 202.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 290DEC
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8948

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12M-708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart