KEY001129474 Key Organics / BIONET

2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD01568852
  • Cas Number : 320423-50-5
  • Molecular Weight : 211.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73 - 75
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/8984

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12N-013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart